Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010231.2 | NP_034361.1 | 3 - 4 | 639 | 73 | |
GenBank mRNA | AK042457.1 | - | 3 - 4 | 486 | 73 | |
AK143940.1 | - | 3 - 4 | 469 | 73 | ||
BC011229.1 | - | 3 - 4 | 639 | 4196446 | 73 | |
D16215.1 | - | 3 - 4 | 529 | 73 | ||
U87456.1 | - | 3 - 4 | 624 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |