Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026250.2 | NP_080526.2 | 6 - 7 | 1083 | 139 | |
GenBank mRNA | AK014287.1 | - | 6 - 7 | 1090 | 139 | |
AK015243.1 | - | 6 - 7 | 1062 | 139 | ||
AK090002.1 | - | 6 - 7 | 801 | 139 | ||
AK169361.1 | - | 6 - 7 | 1046 | 139 | ||
BC033572.1 | - | 5 - 6 | 1028 | 5067692 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |