Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005508.2 | NP_001005508.2 | 3 - 4 | 656 | 96 | |
GenBank mRNA | AK031319.1 | - | 3 - 4 | 427 | 96 | |
AK147757.1 | - | 3 - 4 | 663 | 96 | ||
AK156351.1 | - | 3 - 4 | 645 | 96 | ||
BC082573.1 | - | 3 - 4 | 604 | 30548035 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |