Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153555.2 | NP_705783.1 | 9 - 10 | 1430 | 103 | |
GenBank mRNA | AK035701.1 | - | 9 - 10 | 1329 | 103 | |
AK098114.1 | - | 9 - 10 | 1449 | 103 | ||
AK146825.1 | - | 9 - 10 | 1357 | 103 | ||
AK164799.1 | - | 9 - 10 | 1357 | 103 | ||
AK167059.1 | - | 9 - 10 | 1306 | 103 | ||
BC023804.1 | - | 10 - 11 | 1484 | 5352941 | 103 | |
BC078641.1 | - | 9 - 10 | 2342 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |