Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sctr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012322.2 | NP_001012322.2 | 2 - 3 | 470 | 68 | |
GenBank mRNA | AK032652.1 | - | 2 - 3 | 472 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Mus musculus Probe spans exons |