Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181750.2 | NP_861415.2 | 8 - 9 | 987 | 118 | |
GenBank mRNA | AK141150.1 | - | 6 - 7 | 841 | 118 | |
AK144098.1 | - | 6 - 7 | 861 | 118 | ||
BC049181.1 | - | 8 - 9 | 969 | 118 | ||
BC137772.1 | - | 7 - 8 | 732 | 8860791 | 118 | |
BC144899.1 | - | 7 - 8 | 732 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |