Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011439.2 | NP_035569.2 | 10 - 11 | 1447 | 102 | |
GenBank mRNA | AB006329.1 | - | 9 - 10 | 1103 | 102 | |
AJ000740.1 | - | 9 - 10 | 1054 | 102 | ||
BC007130.1 | - | 10 - 11 | 1269 | 3493465 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |