Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10356680 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10356679 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10356678 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1434823_PM_x_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1433516_PM_a_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1433517_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10356677 |
- |
- |
Mouse Genome 430 2.0 Array |
1433516_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1433517_at |
- |
- |
Mouse Genome 430 2.0 Array |
1434823_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1433517_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1433516_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1434823_PM_x_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
66915_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
66915_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0100022771.mm.1 |
NM_001163425.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022771.mm.1 |
NM_001163425.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022771.mm.1 |
NR_028108.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022771.mm.1 |
NR_028108.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022771.mm.1 |
TC0100002816.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100022772.mm.1 |
NM_001163425.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022772.mm.1 |
NM_001163425.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022772.mm.1 |
NR_028108.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022772.mm.1 |
NR_028108.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100022772.mm.1 |
TC0100002816.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1433517_at |
- |
- |
Mouse Expression Set 430 |
1433516_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1433517_PM_at |
- |
- |
Mouse Expression Set 430 |
1433516_a_at |
- |
- |
Mouse Expression Set 430 |
1434823_x_at |
- |
- |
Mouse Expression Set 430 |
1434823_PM_x_at |
- |
- |
Murine Genome U74v2 Set |
96934_PM_at |
- |
- |
Murine Genome U74v2 Set |
96934_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1434823_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1433516_PM_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1433517_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1433517_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1433516_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1434823_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1433517_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1433516_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1434823_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1433516_PM_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1433517_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1434823_x_at |
- |
- |