Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028942.4 | NP_083218.1 | 7 - 8 | 1372 | 59 | |
GenBank mRNA | AK016647.1 | - | 7 - 8 | 1320 | 59 | |
AK029505.1 | - | 7 - 8 | 1373 | 59 | ||
AK132668.1 | - | 7 - 8 | 1323 | 59 | ||
AK161249.1 | - | 7 - 8 | 1323 | 59 | ||
BC139066.1 | - | 7 - 8 | 1275 | 8862088 | 59 | |
BC139067.1 | - | 7 - 8 | 1275 | 8862089 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |