Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172157.1 | NP_001165628.1 | 1 - 2 | 69 | 69 | |
NM_153530.2 | NP_705758.1 | 1 - 2 | 69 | 69 | ||
GenBank mRNA | AK029652.1 | - | 1 - 2 | 46 | 69 | |
AK031180.1 | - | 1 - 2 | 54 | 69 | ||
AK040613.1 | - | 1 - 2 | 46 | 69 | ||
AK050762.1 | - | 1 - 2 | 70 | 69 | ||
AK079867.1 | - | 1 - 2 | 66 | 69 | ||
BB654969.1 | - | 1 - 2 | 70 | 69 | ||
CJ063044.1 | - | 1 - 2 | 48 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |