Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1456169_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456169_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456169_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
226654_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
226654_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0100013141.mm.1 |
NM_001164525.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013141.mm.1 |
NM_001164525.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013141.mm.1 |
TC0100003871.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1456169_at |
- |
- |
Mouse Expression Set 430 |
1456169_PM_at |
- |
- |
Murine Genome U74v2 Set |
169509_PM_i_at |
- |
- |
Murine Genome U74v2 Set |
169509_i_at |
- |
- |
Murine Genome U74v2 Set |
170927_PM_r_at |
- |
- |
Murine Genome U74v2 Set |
165949_i_at |
- |
- |
Murine Genome U74v2 Set |
170927_r_at |
- |
- |
Murine Genome U74v2 Set |
165949_PM_i_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1456169_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1456169_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1456169_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1456169_PM_at |
- |
- |