Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133664.3 | NP_598425.2 | 3 - 4 | 1441 | 77 | |
GenBank mRNA | AK083526.1 | - | 3 - 4 | 1442 | 77 | |
BC016257.1 | - | 3 - 4 | 1112 | 4912735 | 77 | |
BC025580.1 | - | 3 - 4 | 1345 | 5325039 | 77 | |
BC031131.1 | - | 3 - 4 | 1288 | 4954891 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |