Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053122.4 | NP_444352.2 | 133 | 63 | ||
GenBank mRNA | AF359564.1 | - | 65 | 63 | ||
AK053361.1 | - | 107 | 63 | |||
AK161351.1 | - | 273 | 63 | |||
BC061049.1 | - | 133 | 63 | |||
BC100557.1 | - | 85 | 5057188 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |