Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010103.4 | NP_034233.1 | 2 - 3 | 709 | 62 | |
GenBank mRNA | AK044978.1 | - | 2 - 3 | 568 | 62 | |
AK045088.1 | - | 2 - 3 | 525 | 62 | ||
AK081003.1 | - | 2 - 3 | 541 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |