Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177471.4 | NP_803422.2 | 4 - 5 | 380 | 76 | |
GenBank mRNA | AK008760.1 | - | 4 - 5 | 351 | 76 | |
AK079810.1 | - | 4 - 5 | 381 | 76 | ||
AK135942.1 | - | 4 - 5 | 357 | 76 | ||
AK170686.1 | - | 4 - 5 | 330 | 76 | ||
BC096763.1 | - | 4 - 5 | 338 | 30053993 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |