Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177471.4 | NP_803422.2 | 5 - 6 | 495 | 110 | |
GenBank mRNA | AK008760.1 | - | 5 - 6 | 466 | 110 | |
AK079810.1 | - | 5 - 6 | 496 | 110 | ||
AK135942.1 | - | 5 - 6 | 472 | 110 | ||
AK170686.1 | - | 5 - 6 | 445 | 110 | ||
BC096763.1 | - | 5 - 6 | 453 | 30053993 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |