Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033409.3 | NP_001028581.1 | 11 - 12 | 1210 | 75 | |
GenBank mRNA | AK052873.1 | - | 6 - 7 | 410 | 75 | |
AK085901.1 | - | 11 - 12 | 1168 | 75 | ||
AK137380.1 | - | 11 - 12 | 1211 | 75 | ||
BC141210.1 | - | 11 - 12 | 1162 | 9055822 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |