Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009791.4 | NP_033921.3 | 15 - 16 | 3801 | 103 | |
GenBank mRNA | AF533752.1 | - | 15 - 16 | 3779 | 103 | |
AK052326.1 | - | 14 - 15 | 3890 | 103 | ||
AY971958.1 | - | 15 - 16 | 3642 | 103 | ||
CV556405.1 | - | 3 - 4 | 514 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |