Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009791.4 | NP_033921.3 | 26 - 27 | 9255 | 141 | |
GenBank mRNA | AF062378.1 | - | 9 - 10 | 1512 | 141 | |
AF533752.1 | - | 26 - 27 | 9233 | 141 | ||
AK048277.1 | - | 9 - 10 | 2068 | 141 | ||
AY971958.1 | - | 25 - 26 | 5301 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |