Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300013185.mm.1 |
NM_001083957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013185.mm.1 |
NM_001083957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013185.mm.1 |
NM_009799.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013185.mm.1 |
NM_009799.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013185.mm.1 |
TC0300001667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013192.mm.1 |
NM_009799.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013192.mm.1 |
NM_009799.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013192.mm.1 |
TC0300001667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013184.mm.1 |
NM_001083957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013184.mm.1 |
NM_001083957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013184.mm.1 |
NM_009799.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013184.mm.1 |
NM_009799.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013184.mm.1 |
TC0300001667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013187.mm.1 |
NM_001083957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013187.mm.1 |
NM_001083957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013187.mm.1 |
TC0300001667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013181.mm.1 |
NM_001083957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013181.mm.1 |
NM_001083957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013181.mm.1 |
NM_009799.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013181.mm.1 |
NM_009799.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013181.mm.1 |
TC0300001667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013183.mm.1 |
NM_001083957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013183.mm.1 |
NM_001083957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013183.mm.1 |
NM_009799.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013183.mm.1 |
NM_009799.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013183.mm.1 |
TC0300001667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013186.mm.1 |
NM_001083957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013186.mm.1 |
NM_001083957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013186.mm.1 |
NM_009799.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013186.mm.1 |
NM_009799.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013186.mm.1 |
TC0300001667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013180.mm.1 |
NM_001083957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013180.mm.1 |
NM_001083957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013180.mm.1 |
NM_009799.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013180.mm.1 |
NM_009799.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013180.mm.1 |
TC0300001667.mm.1 |
MTA TC |