Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199213.1 | NP_001186142.1 | 3 - 4 | 368 | 71 | |
NM_027971.2 | NP_082247.1 | 3 - 4 | 438 | 71 | ||
GenBank mRNA | AK009018.1 | - | 3 - 4 | 439 | 71 | |
AK014669.1 | - | 3 - 4 | 371 | 71 | ||
AK040697.1 | - | 3 - 4 | 372 | 71 | ||
AY367773.1 | - | 2 - 3 | 319 | 71 | ||
BC062134.1 | - | 3 - 4 | 343 | 30287757 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |