Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028399.1 | NP_082675.1 | 4 - 5 | 500 | 89 | |
GenBank mRNA | AK013634.1 | - | 4 - 5 | 447 | 89 | |
AK036772.1 | - | 448 | 89 | |||
AK049863.1 | - | 4 - 5 | 445 | 89 | ||
AK154439.1 | - | 4 - 5 | 458 | 89 | ||
AK156231.1 | - | 469 | 89 | |||
BC054122.1 | - | 4 - 5 | 500 | 6509024 | 89 | |
JX965957.1 | - | 4 - 5 | 436 | 89 | ||
JX965958.1 | - | 4 - 5 | 436 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |