Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178912.3 | NP_849243.2 | 20 - 21 | 5323 | 92 | |
GenBank mRNA | AK028941.1 | - | 3 - 4 | 692 | 92 | |
AK162428.1 | - | 1 - 2 | 363 | 92 | ||
AK173211.1 | - | 20 - 21 | 5343 | 92 | ||
BC150785.1 | - | 20 - 21 | 5317 | 9087696 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |