Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207531.3 | NP_997414.2 | 3 - 4 | 308 | 78 | |
GenBank mRNA | AK053177.1 | - | 3 - 4 | 237 | 78 | |
BC023499.1 | - | 3 - 4 | 309 | 5008294 | 78 | |
BE650904.1 | - | 3 - 4 | 308 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |