Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039482.1 | NP_001034571.1 | 7 - 8 | 1314 | 76 | |
GenBank mRNA | AK032131.1 | - | 5 - 6 | 801 | 76 | |
AK053102.1 | - | 7 - 8 | 1264 | 76 | ||
AK054169.1 | - | 7 - 8 | 1282 | 76 | ||
AK220547.1 | - | 7 - 8 | 1293 | 76 | ||
BC019571.1 | - | 7 - 8 | 1286 | 4235202 | 76 | |
BC060183.1 | - | 7 - 8 | 1243 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |