Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025840.3 | NP_080116.2 | 4 - 5 | 511 | 148 | |
GenBank mRNA | AB244988.1 | - | 4 - 5 | 427 | 148 | |
AK003229.1 | - | 4 - 5 | 506 | 148 | ||
AK011253.1 | - | 4 - 5 | 467 | 148 | ||
AK088108.1 | - | 4 - 5 | 443 | 148 | ||
AK132595.1 | - | 4 - 5 | 453 | 148 | ||
AK160544.1 | - | 4 - 5 | 484 | 148 | ||
AK162647.1 | - | 4 - 5 | 453 | 148 | ||
BC013060.1 | - | 4 - 5 | 447 | 3482251 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |