Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027117.3 | NP_081393.2 | 8 | 1051 | 94 | |
GenBank mRNA | AK009481.1 | - | 8 | 966 | 94 | |
AY513272.1 | - | 8 | 1002 | 94 | ||
BC005581.1 | - | 8 | 975 | 3968615 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Amplicon spans exons and probe does not span exons |