Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029860.1 | - | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Probe spans exons |