Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080793.5 | NP_542983.3 | 4 - 5 | 832 | 72 | |
GenBank mRNA | AF448509.1 | - | 4 - 5 | 614 | 72 | |
AK048924.1 | - | 4 - 5 | 669 | 72 | ||
AK129428.1 | - | 4 - 5 | 621 | 72 | ||
AK147316.1 | - | 4 - 5 | 888 | 72 | ||
AK147413.1 | - | 4 - 5 | 658 | 72 | ||
AK147422.1 | - | 4 - 5 | 833 | 72 | ||
AK147667.1 | - | 4 - 5 | 867 | 72 | ||
AK170161.1 | - | 4 - 5 | 676 | 72 | ||
BC050190.1 | - | 4 - 5 | 656 | 30007831 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |