Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013529.3 | NP_038557.1 | 10 - 11 | 1046 | 81 | |
GenBank mRNA | AB016780.1 | - | 10 - 11 | 1012 | 81 | |
AK132821.1 | - | 9 - 10 | 892 | 81 | ||
BC031928.1 | - | 10 - 11 | 1014 | 4167189 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |