Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013529.3 | NP_038557.1 | 11 - 12 | 1134 | 72 | |
GenBank mRNA | AB016780.1 | - | 11 - 12 | 1100 | 72 | |
AK132821.1 | - | 10 - 11 | 980 | 72 | ||
BC031928.1 | - | 11 - 12 | 1102 | 4167189 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |