Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100012121.mm.1 |
NM_001039045.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012121.mm.1 |
NM_001039045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012121.mm.1 |
NM_026078.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012121.mm.1 |
NM_026078.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012121.mm.1 |
TC0100001473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012124.mm.1 |
NM_001039045.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012124.mm.1 |
NM_001039045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012124.mm.1 |
NM_026078.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012124.mm.1 |
NM_026078.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012124.mm.1 |
TC0100001473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012125.mm.1 |
NM_001039045.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012125.mm.1 |
NM_001039045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012125.mm.1 |
NM_026078.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012125.mm.1 |
NM_026078.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012125.mm.1 |
TC0100001473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012123.mm.1 |
NM_001039045.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012123.mm.1 |
NM_001039045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012123.mm.1 |
NM_026078.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012123.mm.1 |
NM_026078.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012123.mm.1 |
TC0100001473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012122.mm.1 |
NM_001039045.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012122.mm.1 |
NM_001039045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012122.mm.1 |
NM_026078.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012122.mm.1 |
NM_026078.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012122.mm.1 |
TC0100001473.mm.1 |
MTA TC |