Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033156.4 | NP_001028328.2 | 1 - 2 | 626 | 109 | |
GenBank mRNA | BC020022.1 | - | 1 - 2 | 595 | 109 | |
BC025217.1 | - | 1 - 2 | 206 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |