Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081281.1 | NP_001074750.1 | 4 - 5 | 751 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |