Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011158.3 | NP_035288.2 | 4 - 5 | 673 | 96 | |
GenBank mRNA | AK011647.1 | - | 4 - 5 | 631 | 96 | |
AK041013.1 | - | 4 - 5 | 647 | 96 | ||
AK081805.1 | - | 4 - 5 | 450 | 96 | ||
AK135621.1 | - | 4 - 5 | 675 | 96 | ||
AK138963.1 | - | 4 - 5 | 639 | 96 | ||
BC048710.1 | - | 4 - 5 | 629 | 6506827 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |