Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081125.1 | NP_001074594.1 | 12 - 13 | 2255 | 84 | |
GenBank mRNA | BC082604.1 | - | 12 - 13 | 2196 | 30672479 | 84 |
BC085190.1 | - | 12 - 13 | 2171 | 30694633 | 84 | |
X99104.1 | - | 12 - 13 | 2015 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |