Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mael | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175296.4 | NP_780505.1 | 4 - 5 | 564 | 82 | |
GenBank mRNA | AK077103.1 | - | 4 - 5 | 561 | 82 | |
AK165801.1 | - | 4 - 5 | 561 | 82 | ||
BC050800.1 | - | 4 - 5 | 564 | 6744032 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |