Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011804.2 | NP_035934.1 | 1 - 2 | 359 | 154 | |
GenBank mRNA | AF084524.1 | - | 1 - 2 | 359 | 154 | |
AK005388.1 | - | 1 - 2 | 380 | 154 | ||
AK010947.1 | - | 1 - 2 | 376 | 154 | ||
AK085844.1 | - | 1 - 2 | 171 | 154 | ||
AK144101.1 | - | 1 - 2 | 378 | 154 | ||
AK191906.1 | - | 1 - 2 | 68 | 154 | ||
AK200846.1 | - | 1 - 2 | 68 | 154 | ||
AK203453.1 | - | 1 - 2 | 68 | 154 | ||
AK204882.1 | - | 1 - 2 | 154 | |||
BC027426.1 | - | 1 - 2 | 359 | 3670649 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |