Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007955.3 | NP_031981.3 | 28 - 29 | 5023 | 55 | |
GenBank mRNA | AF300701.1 | - | 27 - 28 | 4500 | 55 | |
AK156112.1 | - | 28 - 29 | 5026 | 55 | ||
U36488.1 | - | 24 - 25 | 4185 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Detect Genomic DNA Mus musculus Probe spans exons |