Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK147106.1 | - | 1 - 2 | 1805 | 76 | |
AK162481.1 | - | 1 - 2 | 1513 | 76 | ||
AK220375.1 | - | 1 - 2 | 1802 | 76 | ||
BC060298.1 | - | 1 - 2 | 597 | 76 | ||
BC145902.1 | - | 1 - 2 | 1484 | 40131205 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |