Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033300.2 | NP_001028472.2 | 10 - 11 | 1454 | 110 | |
GenBank mRNA | AK049755.1 | - | 10 - 11 | 1418 | 110 | |
AK143197.1 | - | 10 - 11 | 1455 | 110 | ||
AK146654.1 | - | 10 - 11 | 1472 | 110 | ||
AK167857.1 | - | 10 - 11 | 1431 | 110 | ||
AK168239.1 | - | 10 - 11 | 1404 | 110 | ||
AK169043.1 | - | 10 - 11 | 1438 | 110 | ||
AK169701.1 | - | 10 - 11 | 1452 | 110 | ||
BC037232.1 | - | 8 - 9 | 1106 | 110 | ||
BC080685.1 | - | 5 - 6 | 666 | 110 | ||
BC138411.1 | - | 10 - 11 | 1410 | 8861431 | 110 | |
BC138412.1 | - | 10 - 11 | 1410 | 8861432 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |