Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081406.1 | NP_001074875.1 | 2 - 3 | 353 | 78 | |
GenBank mRNA | AK144741.1 | - | 2 - 3 | 354 | 78 | |
BC075711.1 | - | 2 - 3 | 335 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |