Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172845.2 | NP_766433.1 | 3 - 4 | 1502 | 72 | |
GenBank mRNA | AK028364.1 | - | 1503 | 72 | ||
AK076295.1 | - | 2 - 3 | 877 | 72 | ||
AK083534.1 | - | 3 - 4 | 1502 | 72 | ||
AK156429.1 | - | 1 - 2 | 85 | 72 | ||
AK157287.1 | - | 1503 | 72 | |||
AK164900.1 | - | 3 - 4 | 1498 | 72 | ||
AK173001.1 | - | 1380 | 72 | |||
BC027773.1 | - | 3 - 4 | 1136 | 5345809 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |