Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100013073.mm.1 |
NM_001122680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013073.mm.1 |
NM_001122680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013073.mm.1 |
NM_027893.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013073.mm.1 |
NM_027893.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013073.mm.1 |
TC0100001589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100013074.mm.1 |
NM_001122680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013074.mm.1 |
NM_001122680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013074.mm.1 |
NM_027893.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013074.mm.1 |
NM_027893.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013074.mm.1 |
TC0100001589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100013070.mm.1 |
NM_001122680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013070.mm.1 |
NM_001122680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013070.mm.1 |
NM_027893.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013070.mm.1 |
NM_027893.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013070.mm.1 |
TC0100001589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100013071.mm.1 |
NM_001122680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013071.mm.1 |
NM_001122680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013071.mm.1 |
NM_027893.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013071.mm.1 |
NM_027893.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013071.mm.1 |
TC0100001589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100013072.mm.1 |
NM_001122680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013072.mm.1 |
NM_001122680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013072.mm.1 |
NM_027893.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013072.mm.1 |
NM_027893.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013072.mm.1 |
TC0100001589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100013067.mm.1 |
NM_001122680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013067.mm.1 |
NM_001122680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013067.mm.1 |
NM_027893.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013067.mm.1 |
NM_027893.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013067.mm.1 |
TC0100001589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100013068.mm.1 |
NM_001122680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013068.mm.1 |
NM_001122680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013068.mm.1 |
NM_027893.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013068.mm.1 |
NM_027893.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100013068.mm.1 |
TC0100001589.mm.1 |
MTA TC |