Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081304.1 | NP_001074773.1 | 3 - 4 | 285 | 87 | |
GenBank mRNA | AK020270.1 | - | 3 - 4 | 265 | 87 | |
AK036106.1 | - | 3 - 4 | 265 | 87 | ||
AK036335.1 | - | 3 - 4 | 283 | 87 | ||
AK087000.1 | - | 3 - 4 | 266 | 87 | ||
BC043662.1 | - | 2 - 3 | 132 | 87 | ||
BC141028.1 | - | 3 - 4 | 279 | 9055640 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |