Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028759.1 | NP_083035.1 | 5 - 6 | 781 | 68 | |
GenBank mRNA | AK004618.1 | - | 5 - 6 | 644 | 68 | |
BC058980.1 | - | 3 - 4 | 344 | 68 | ||
BC084732.1 | - | 3 - 4 | 344 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |