Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028759.1 | NP_083035.1 | 8 - 9 | 1240 | 70 | |
GenBank mRNA | AK004618.1 | - | 8 - 9 | 1103 | 70 | |
BC058980.1 | - | 6 - 7 | 803 | 70 | ||
BC084732.1 | - | 6 - 7 | 803 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |