Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025819.4 | NP_080095.3 | 1 - 2 | 136 | 143 | |
GenBank mRNA | AK013686.1 | - | 1 - 2 | 81 | 143 | |
BC055064.1 | - | 1 - 2 | 6528686 | 143 | ||
BC060204.1 | - | 1 - 2 | 76 | 6855442 | 143 | |
BY062718.1 | - | 1 - 2 | 136 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |