Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039511.1 | NP_001034600.1 | 7 - 8 | 1151 | 92 | |
GenBank mRNA | AB055738.1 | - | 7 - 8 | 1147 | 91 | |
AK007291.1 | - | 7 - 8 | 1101 | 92 | ||
BC040250.1 | - | 7 - 8 | 1143 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |