Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253682.1 | NP_001240611.1 | 8 - 9 | 2311 | 84 | |
NM_016984.3 | NP_058680.1 | 8 - 9 | 2311 | 84 | ||
NR_046161.1 | 8 - 9 | 2258 | 84 | |||
GenBank mRNA | AF011543.1 | - | 8 - 9 | 2128 | 84 | |
AF019663.1 | - | 8 - 9 | 2128 | 84 | ||
AF190646.1 | - | 8 - 9 | 2202 | 84 | ||
AK044265.1 | - | 8 - 9 | 2167 | 84 | ||
AK080670.1 | - | 8 - 9 | 2312 | 84 | ||
BC057335.1 | - | 8 - 9 | 2147 | 84 | ||
BC060160.1 | - | 8 - 9 | 2147 | 84 | ||
BC120729.1 | - | 8 - 9 | 2226 | 40129652 | 84 | |
BC131915.1 | - | 8 - 9 | 2226 | 40130192 | 84 | |
U50921.1 | - | 8 - 9 | 2264 | 84 | ||
U50922.1 | - | 8 - 9 | 2264 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |